COMH04235A44 Combi-Blocks, Inc.

2-(2,5-Dimethoxyphenyl)quinoline-4-carbonyl chloride

  • MDL number MFCD12197775
  • Cas Number : 1160264-87-8
  • Molecular Weight : 327.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4242
Availability details for
Quantity
  • Availability
  • Add to Cart