COMH04235A49 Combi-Blocks, Inc.

6-Chloro-n,n'-bis(3,4-dichlorophenyl)-1,3,5-triazine-2,4-diamine

  • MDL number MFCD01953481
  • Cas Number : 30355-06-7
  • Molecular Weight : 435.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4247
Availability details for
Quantity
  • Availability
  • Add to Cart