COMH04235A58 Combi-Blocks, Inc.

2-Morpholin-2-yl-1h-pyrrolo[2,3-b]pyridine dihydrochloride

  • MDL number MFCD29035004
  • Cas Number : 1503503-78-3
  • Molecular Weight : 276.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4262

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4262
Availability details for
Quantity
  • Availability
  • Add to Cart