COMH04235A5F Combi-Blocks, Inc.

5-Bromo-1-(phenylsulfonyl)indoline

  • MDL number MFCD03368333
  • Cas Number : 118757-04-3
  • Molecular Weight : 338.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4269
Availability details for
Quantity
  • Availability
  • Add to Cart