COMH04235B04 Combi-Blocks, Inc.

N-(Methoxyacetyl)-2-methylalanine

  • MDL number MFCD13559691
  • Cas Number : 1220037-42-2
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4435
Availability details for
Quantity
  • Availability
  • Add to Cart