COMH04235B07 Combi-Blocks, Inc.

N-(4-Acetylphenyl)-2-(4-chloro-2-methylphenoxy)propanamide

  • MDL number MFCD01034770
  • Cas Number : 14157-43-8
  • Molecular Weight : 331.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4438

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4438
Availability details for
Quantity
  • Availability
  • Add to Cart