COMH04235B1A Combi-Blocks, Inc.

2-Propylquinoline-4-carboxylic acid

  • MDL number MFCD01325647
  • Cas Number : 1019-03-0
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4457
Availability details for
Quantity
  • Availability
  • Add to Cart