COMH04235B33 Combi-Blocks, Inc.

4-Amino-6-phenyl-1,6-dihydro-1,3,5-triazine-2-thiol

  • MDL number MFCD12027917
  • Cas Number : 117411-02-6
  • Molecular Weight : 206.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4482
Availability details for
Quantity
  • Availability
  • Add to Cart