COMH04235B34 Combi-Blocks, Inc.

tert-Butyl 4-benzyl-3,5-dioxopiperazine-1-carboxylate

  • MDL number MFCD24849136
  • Cas Number : 1437312-02-1
  • Molecular Weight : 304.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4483
Availability details for
Quantity
  • Availability
  • Add to Cart