COMH04235BBF Combi-Blocks, Inc.

1-(Pyridin-3-ylmethyl)-1h-indole-6-carbaldehyde

  • MDL number MFCD12026989
  • Cas Number : 1030429-74-3
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4622

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4622
Availability details for
Quantity
  • Availability
  • Add to Cart