COMH04235BDA Combi-Blocks, Inc.

6,6-Dimethyl-1-(2-methylphenyl)-4,5,6,7-tetrahydro-1h-indazol-4-amine hydrochloride

  • MDL number MFCD15146558
  • Cas Number : 1242339-61-2
  • Molecular Weight : 291.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4649
Availability details for
Quantity
  • Availability
  • Add to Cart