COMH04235BDE Combi-Blocks, Inc.

2-(2-Piperazin-1-ylethyl)-3a,4,7,7a-tetrahydro-1h-4,7-methanoisoindole-1,3-dione dihydrochloride

  • MDL number MFCD03368385
  • Cas Number : 136369-09-0
  • Molecular Weight : 348.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4653

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4653
Availability details for
Quantity
  • Availability
  • Add to Cart