COMH04235BE7 Combi-Blocks, Inc.

N-(3,4-Dimethoxyphenyl)-n-(phenylsulfonyl)glycine

  • MDL number MFCD02576287
  • Cas Number : 333449-93-7
  • Molecular Weight : 351.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4662
Availability details for
Quantity
  • Availability
  • Add to Cart