COMH04235C29 Combi-Blocks, Inc.

Ethyl oxo(piperidin-2-yl)acetate hydrochloride

  • MDL number MFCD28142419
  • Cas Number : 1823547-10-9
  • Molecular Weight : 221.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4728
Availability details for
Quantity
  • Availability
  • Add to Cart