COMH04235C37 Combi-Blocks, Inc.

2-(2-Aminoethyl)-1,3-benzoxazole-5-sulfonamide

  • MDL number MFCD27995995
  • Cas Number : 1785761-66-1
  • Molecular Weight : 241.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4742
Availability details for
Quantity
  • Availability
  • Add to Cart