COMH04235C69 Combi-Blocks, Inc.

Ethyl 2-(4-aminophenoxy)isonicotinate

  • MDL number MFCD22689493
  • Cas Number : 1415719-23-1
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4792
Availability details for
Quantity
  • Availability
  • Add to Cart