COMH04235C83 Combi-Blocks, Inc.

N-[(2-Methyl-1,3-thiazol-4-yl)methyl]-l-prolinamide hydrochloride

  • MDL number MFCD22123358
  • Cas Number : 1417575-71-3
  • Molecular Weight : 261.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4818
Availability details for
Quantity
  • Availability
  • Add to Cart