COMH04235C86 Combi-Blocks, Inc.

3-(1H-Benzimidazol-2-yl)propanimidamide

  • MDL number MFCD19103474
  • Cas Number : 1306739-41-2
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4821
Availability details for
Quantity
  • Availability
  • Add to Cart