COMH04235CA0 Combi-Blocks, Inc.

2-[5-(4-Fluorobenzyl)-1,3-oxazol-2-yl]piperidine dihydrochloride

  • MDL number MFCD27756605
  • Cas Number : 1982950-51-5
  • Molecular Weight : 333.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4847

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4847
Availability details for
Quantity
  • Availability
  • Add to Cart