COMH04235D09 Combi-Blocks, Inc.

Methyl (5-oxo-4,5,6,7-tetrahydro[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)acetate

  • MDL number MFCD11058053
  • Cas Number : 1030575-22-4
  • Molecular Weight : 210.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4952
Availability details for
Quantity
  • Availability
  • Add to Cart