COMH04235D15 Combi-Blocks, Inc.

Methyl 3-([(1e)-(dimethylamino)methylene]amino)-5-methyl-1h-indole-2-carboxylate

  • MDL number MFCD15146535
  • Cas Number : 1217885-74-9
  • Molecular Weight : 259.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-4964
Availability details for
Quantity
  • Availability
  • Add to Cart