COMH04235D7B Combi-Blocks, Inc.

3-[5-(4-Fluorobenzyl)-1,3-oxazol-2-yl]piperidine dihydrochloride

  • MDL number MFCD27756599
  • Cas Number : 1982950-43-5
  • Molecular Weight : 333.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5066
Availability details for
Quantity
  • Availability
  • Add to Cart