COMH04235D9B Combi-Blocks, Inc.

4-[(2-Fluorophenyl)(methylsulfonyl)amino]butanoic acid

  • MDL number MFCD28954540
  • Cas Number : 1266545-23-6
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5098
Availability details for
Quantity
  • Availability
  • Add to Cart