COMH04235E1B Combi-Blocks, Inc.

4-Methyl-2-(methylamino)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD02855638
  • Cas Number : 110859-69-3
  • Molecular Weight : 172.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5227
Availability details for
Quantity
  • Availability
  • Add to Cart