COMH04235E23 Combi-Blocks, Inc.

4-[(E)-2-(3-Fluorophenyl)vinyl]pyridine hydrochloride

  • MDL number MFCD12028059
  • Cas Number : 1185236-01-4
  • Molecular Weight : 235.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5235

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5235
Availability details for
Quantity
  • Availability
  • Add to Cart