COMH04235E66 Combi-Blocks, Inc.

[2-(2-Ethylimidazo[2,1-b][1,3,4]thiadiazol-6-yl)ethyl]amine dihydrochloride

  • MDL number MFCD18071418
  • Cas Number : 1332528-50-3
  • Molecular Weight : 269.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5303

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5303
Availability details for
Quantity
  • Availability
  • Add to Cart