COMH04235EE6 Combi-Blocks, Inc.

1-Isopropyl-3-methyl-1h-pyrazole-4-carbaldehyde

  • MDL number MFCD06805437
  • Cas Number : 1006453-59-3
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5431
Availability details for
Quantity
  • Availability
  • Add to Cart