COMH04235F74 Combi-Blocks, Inc.

1-Benzyl-1h-imidazole-2-carbaldehyde

  • MDL number MFCD00088667
  • Cas Number : 10045-65-5
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5573

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5573
Availability details for
Quantity
  • Availability
  • Add to Cart