COMH04235F9A Combi-Blocks, Inc.

[5-(Chloromethyl)-2-furyl](4-chlorophenyl)methanone

  • MDL number MFCD05860749
  • Cas Number : 2856-02-2
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5611
Availability details for
Quantity
  • Availability
  • Add to Cart