COMH04235FD6 Combi-Blocks, Inc.

5-(2-Furyl)-2-methylthieno[2,3-d]pyrimidin-4(3h)-one

  • MDL number MFCD11569321
  • Cas Number : 1082134-24-4
  • Molecular Weight : 232.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5672
Availability details for
Quantity
  • Availability
  • Add to Cart