COMH04236029 Combi-Blocks, Inc.

Methyl 3-(4-aminophenyl)-2-phenylpropanoate hydrochloride

  • MDL number MFCD27756545
  • Cas Number : 1211569-04-8
  • Molecular Weight : 291.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5755
Availability details for
Quantity
  • Availability
  • Add to Cart