COMH0423603A Combi-Blocks, Inc.

Ethyl 2-(4-nitro-1h-pyrazol-1-yl)butanoate

  • MDL number MFCD04969812
  • Cas Number : 1005668-54-1
  • Molecular Weight : 227.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5772
Availability details for
Quantity
  • Availability
  • Add to Cart