COMH04236087 Combi-Blocks, Inc.

1-Butyrylpiperidine-4-carboxylic acid

  • MDL number MFCD09816262
  • Cas Number : 117704-87-7
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5849

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5849
Availability details for
Quantity
  • Availability
  • Add to Cart