COMH042360B9 Combi-Blocks, Inc.

Ethyl 4-(3-[(cyclopropylmethyl)amino]-2,5-dioxopyrrolidin-1-yl)benzoate

  • MDL number MFCD22689490
  • Cas Number : 1415719-11-7
  • Molecular Weight : 316.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-5899
Availability details for
Quantity
  • Availability
  • Add to Cart