COMH04236122 Combi-Blocks, Inc.

7-(2-Hydroxyethyl)-9-phenylpyrido[4,3-d][1,2,4]triazolo[1,5-a]pyrimidin-8(7h)-one

  • MDL number MFCD13872802
  • Cas Number : 1158403-10-1
  • Molecular Weight : 307.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6004
Availability details for
Quantity
  • Availability
  • Add to Cart