COMH0423615B Combi-Blocks, Inc.

[1-(2-Thienyl)cyclopropyl]amine hydrochloride

  • MDL number MFCD19103326
  • Cas Number : 1332530-75-2
  • Molecular Weight : 175.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6063

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6063
Availability details for
Quantity
  • Availability
  • Add to Cart