COMH04236175 Combi-Blocks, Inc.

2-Hydroxy-5-(pyrrolidin-1-ylsulfonyl)benzoic acid

  • MDL number MFCD02700702
  • Cas Number : 326024-05-9
  • Molecular Weight : 271.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6089

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6089
Availability details for
Quantity
  • Availability
  • Add to Cart