COMH042361B1 Combi-Blocks, Inc.

2,3-Diphenylcycloprop-2-ene-1-carboxylic acid

  • MDL number MFCD00157043
  • Cas Number : 17825-58-0
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6149
Availability details for
Quantity
  • Availability
  • Add to Cart