COMH042361C9 Combi-Blocks, Inc.

1-[(2,4-Dimethylphenoxy)methyl]-1h-pyrazol-4-amine hydrochloride

  • MDL number MFCD09431206
  • Cas Number : 1049764-63-7
  • Molecular Weight : 253.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6173

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6173
Availability details for
Quantity
  • Availability
  • Add to Cart