COMH042361EA Combi-Blocks, Inc.

1-(3-Methyl-1,2,4-oxadiazol-5-yl)ethan-1-amine hydrochloride

  • MDL number MFCD13857386
  • Cas Number : 1041578-67-9
  • Molecular Weight : 163.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6206
Availability details for
Quantity
  • Availability
  • Add to Cart