COMH042361FF Combi-Blocks, Inc.

tert-Butyl [(1e)-amino(1h-pyrazol-1-yl)methylene]carbamate

  • MDL number MFCD22374938
  • Cas Number : 152120-61-1
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6227
Availability details for
Quantity
  • Availability
  • Add to Cart