COMH0423626F Combi-Blocks, Inc.

N-Allyl-2-(3-chlorobenzoyl)hydrazinecarbothioamide

  • MDL number MFCD01907094
  • Cas Number : 15944-95-3
  • Molecular Weight : 269.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6339

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6339
Availability details for
Quantity
  • Availability
  • Add to Cart