COMH0423635E Combi-Blocks, Inc.

2-Cyclopropyl-1,3-benzothiazole

  • MDL number MFCD28505144
  • Cas Number : 1248473-57-5
  • Molecular Weight : 175.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6580

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6580
Availability details for
Quantity
  • Availability
  • Add to Cart