COMH04236391 Combi-Blocks, Inc.

Methyl 2-[(2,3-dimethylphenyl)amino]benzoate

  • MDL number MFCD01661278
  • Cas Number : 1222-42-0
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6631

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6631
Availability details for
Quantity
  • Availability
  • Add to Cart