COMH042363A8 Combi-Blocks, Inc.

N,N-Dimethyl-3-(6-pyrrolidin-2-ylpyridin-2-yl)propanamide dihydrochloride

  • MDL number MFCD29035061
  • Cas Number : 1858251-53-2
  • Molecular Weight : 320.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6654
Availability details for
Quantity
  • Availability
  • Add to Cart