COMH042363B0 Combi-Blocks, Inc.

1H-Indol-3-yl[(3-methylbutyl)amino]acetic acid

  • MDL number MFCD13176345
  • Cas Number : 1214860-66-8
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6662
Availability details for
Quantity
  • Availability
  • Add to Cart