COMH042363EA Combi-Blocks, Inc.

3-Chloro-n-(2,2-dimethoxyethyl)pyrazin-2-amine

  • MDL number MFCD24849105
  • Cas Number : 1417361-15-9
  • Molecular Weight : 217.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6720
Availability details for
Quantity
  • Availability
  • Add to Cart