COMH042363F1 Combi-Blocks, Inc.

3-Methoxy-4-thiomorpholin-4-ylcyclobut-3-ene-1,2-dione

  • MDL number MFCD28954480
  • Cas Number : 1858251-77-0
  • Molecular Weight : 213.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6727
Availability details for
Quantity
  • Availability
  • Add to Cart