COMH042363FB Combi-Blocks, Inc.

4-(2-Aminoethoxy)-3-methoxybenzoic acid hydrochloride

  • MDL number MFCD16617506
  • Cas Number : 1011408-00-6
  • Molecular Weight : 247.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6737

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6737
Availability details for
Quantity
  • Availability
  • Add to Cart