COMH0423640C Combi-Blocks, Inc.

Ethyl 2-(5-methyl-3-oxopiperazin-2-yl)acetate

  • MDL number MFCD18157633
  • Cas Number : 5194-01-4
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6754
Availability details for
Quantity
  • Availability
  • Add to Cart